missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well Clear Polystyrene Microplates

Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 384-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.
£682.00 - £747.27
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Color | Clear |
Material | Polystyrene |
No. of Wells | 384 |
Product Code | Brand | Surface Treatment | Lid | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Lid | Price | Quantity & Availability | |||||
10134431
![]() |
Corning™
3700 |
High Binding | Without Lid |
£736.12
Pack of 100 |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10481242
![]() |
Corning™
3702 |
Non-treated | Without Lid |
£682.00
Pack of 100 |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
10544961
![]() |
Corning™
3680 |
Non-treated | With Lid |
£747.27
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
Specifications
Standard | |
Clear | |
384 | |
Flat Bottom (F-well) | |
100/Cs. |
16 x 24 | |
Polystyrene | |
20 to 80 μL | |
100/Cs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 384-Well Clear Polystyrene Microplates