Learn More
Corning™ Reusable Media Handling Fittings
Introduce medium aseptically into larger Spinner Flasks with vertical sidearms during cell culture
Brand:  Corning™ 4519-120
 View more versions of this product
Product Code. 10275982
  • £622.06 / Each
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Used to introduce medium aseptically into large spinner flasks with vertical sidearms.
  • Fittings are comprised of a PET insert with a fluorocarbon O-Ring and a polypropylene sealing cap.
  • Gas filters are PTFE, 0.2 μm porosity.
  • The 316 stainless steel tubes are held in place by modified polyphenylene oxide thermoplastic nuts with integrated ferrules.
  • The fittings are completely autoclavable.
Spare parts for sidearm fittings, see Cat. Nos. 1395-32LTC, 1395-45LTC, 1395-LTR, and 1395LTMC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Orange
2 tubes; Tube style: Straight
0.31, 0.63 cm
PET insert, Viton O-rings, 45 mm Polypropylene sealing cap, Stainless Steel tubes, Noryl nuts, integrated ferrules
1/Pk.
GL 45
Reusable
Screw Cap
Polypropylene
1/8 and 1/4 in.
8 or 15 L Vertical Sidearm Flask
1/Cs.
Media handling fitting
Autoclavable
Non-sterile
45 mm (approx.)