missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Sterile Spatulas, Tapered blade/spoon
BIOPHARMA

Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.

Brand:  Corning™ 3003

Product Code. 10788012

  • £256.00 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.

  • Corning spatulas are designed to save time and to provide contamination-free samples.
  • Eliminates the recycling and resterilizing necessary with reusable spatulas.
  • Available in five different configurations.
  • Microspatulas available in two configurations.
  • Individually packaged.
  • Antistatic.
  • Sterile.
  • RNase-/DNase-free.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sterile
USP Class VI, non-pyrogenic, RNase and DNase-free Certified
Spatula
9.8 in.
Polystyrene
24.9 cm
Tapered Flat End, Spoon End
Single-use
Molecular biology applications, lyophilized samples and contamination-free sampling
Tapered flat blade at one end and a spoon at the other end
Tapered
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number31624038 Certificate TypeCertificate of Quality Date22/02/2025 Catalog Number3003
Lot Number19124054 Certificate TypeCertificate of Quality Date25/12/2024 Catalog Number3003
Lot Number14224067 Certificate TypeCertificate of Quality Date27/07/2024 Catalog Number3003
Lot Number19024084 Certificate TypeCertificate of Compliance Date03/09/2024 Catalog Number3003
Lot Number14224067 Certificate TypeCertificate of Compliance Date17/07/2024 Catalog Number3003
Special Offers

Special Offers

  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Sterile Spatulas, Tapered blade/spoon > Spatula, Tapered Blade/Spoon

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.