Learn More
Corning™ Surface Modified Stripwell™ Microplates
Four surface chemistries suit every type of assay
Brand:  Corning™ 2506
Code : 67
Additional Details : Weight : 0.02385kg
 View more versions of this product
Product Code. 10756482
  • £733.95 / Pack of 50
Estimated Shipment: 15-10-2024
to see stock.
Add to basket
Description
Description
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format.
  • Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
  • 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Specifications
Specifications
Strip Plate
8 x 12
Clear
96
Non-sterile
360 μL
In-vitro diagnostic assays
1/Pk.
Without Lid
DNA-Bind
Polystyrene
75 to 200 μL
DNA-Bind
Round
50/Cs.
50/Cs.