missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Surface Modified Stripwell™ Microplates
Four surface chemistries suit every type of assay
£733.95
Specifications
Format | Strip Plate |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
Color | Clear |
Material | Polystyrene |
Description
Corning™ Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an “egg-crate” style strip holder that allows each individual well to be positioned back into the microplate once broken.
- Stripwell microplates have 96-well flat bottom polystyrene format.
- Low volume and standard Stripwell microplates have well volumes of 190 μL and 360 μL, respectively.
- 1 x 8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation.
Specifications
Strip Plate | |
8 x 12 | |
Polystyrene | |
75 to 200 μL | |
360 μL | |
50/Cs. |
Without Lid | |
Clear | |
96 | |
Non-sterile | |
In-vitro diagnostic assays |