missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Thermowell™ GOLD Polypropylene PCR Tubes

Designed for precise fit in heat blocks

Brand:  Corning™ 3746

Code : 66

Additional Details : Weight : 0.22680kg

 View more versions of this product

Product Code. 10511503

  • £114.17 / Pack of 1000
In Stock
to see stock.

Description

Description

Individual PCR tubes are made of thin wall polypropylene and designed for precise fit in heat blocks to optimize heat transfer. Tubes are RNase-/DNase-free, are autoclavable at 121°C, and withstand centrifugation to 10,000 x g.

Specifications

Specifications

RNase, DNase-free
Assorted
500/Pk.
Heat blocks to optimize heat transfer
Non-sterile
10,000 x g
Autoclavable
Thin Wall PCR Tube
Assorted
0.2 mL
Polypropylene
Flat
Polypropylene
Domed
Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers