missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Universal CellSTACK™ Caps
GREENER_CHOICE

For filling, venting, and manipulation

Brand:  Corning™ 3328

 View more versions of this product

Product Code. 10457163

  • £282.00 / Pack of 4
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Choice of 33mm polyethylene, solid or vented
  • Sterilized by gamma irradiation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orange
3.3 cm
Female MPC Polycarbonate with a 0.25 in. I.D. coupling and a male MPC Polycarbonate end cap
Single-use
Screw
HDPE
For filling, venting, and manipulation
Cap, Filling
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.