Learn More
Corning™ Universal Fit Pipet Tips: Bulk Packed
Designed to assure a reliable fit with major brand pipetters
Brand: Corning™ 4845
Code : 66
Additional Details : Weight : 3.14340kg
Description
Designed to assure a reliable fit with major brand pipetters
- Beveled orifice helps ensure accurate fluid delivery
- 1–200μL tips are graduated at 10, 50 and 100μL
- 100–1000μL tips are graduated at 250, 500, and 1000μL
- Choice of sterile or nonsterile
- Bulk packed tips are nonsterile and very economical
Specifications
1 to 200 μL | |
Yellow | |
Round | |
Yes |
Non-sterile | |
Bulk | |
Universal Fit | |
Polypropylene |