Learn More
Corning™ Universal Fit Pipet Tips: Racked, Nonsterile
Designed to assure a reliable fit with major brand pipetters
Brand: Corning™ 4865
57.81 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
Designed to assure a reliable fit with major brand pipetters
- Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
- Beveled orifice helps ensure accurate fluid delivery.
- 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
- Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
1 to 200 μL | |
Non-sterile | |
Rack | |
Rack | |
Yes |
Non-filtered | |
Yellow | |
Round | |
Universal Fit | |
Polypropylene |