Learn More
Invitrogen™ Corticosterone Competitive ELISA Kit
Competitive ELISA
£534.00 - £3590.00
Specifications
Assay Sensitivity | 18.6 pg/mL |
---|---|
Description | Corticosterone Competitive ELISA Kit |
Conjugate | HRP |
Detection Method | Fluorescence |
Product Type | ELISA Kit |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
15805901
|
Invitrogen™
EIACORT |
96 assays |
£534.00
Pack of 96 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17845117
|
Invitrogen™
EIACORTX10 |
10 x 96 Assays |
£3590.00
Pack of 10 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
Description
The Corticosterone ELISA quantitates coritcosterone in serum, plasma, urine, dried fecal extracts or cell culture medium. The assay will exclusively recognize both natural and recombinant coritcosterone. Principle of the method The corticosterone solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is inversely proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CORTICOSTERONE is an adrenocortical steroid that has modest but significant activities as a mineralocorticoid and a glucocorticoid. [National Center for Biotechnology Information. PubChem Compound Database; CID=5753].Specifications
18.6 pg/mL | |
HRP | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
5.2% | |
RUO | |
-20°C | |
1 hr. 20 min. |
Corticosterone Competitive ELISA Kit | |
Fluorescence | |
Fecal Extract, Plasma, Serum, Supernatant, Urine | |
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti | |
7.9% | |
HRP | |
Inflammation | |
Chemical | |
1 hr. 30 min. |