Learn More
Corning™ Costar™ 8-Pette™ and 12-Pette™ Multichannel Pipetters
Adjustable volume pipetters can use any number of channels without affecting pipetting accuracy
Brand: Corning™ 4888
This item is not returnable.
View return policy
Description
These20-200 μL multi-channel pipettors feature a unique, ergonomic trigger-style aspiration and dispense control mechanism designed to reduce thumb fatigue during repetitive pipetting.
• Volume adjusts with a vernier-scale spindle
• Pipettors are entirely autoclavable
Order Info
Compatible with all Fisherbrand™ universal-fit tips.
Specifications
Autoclavable | |
Costar™ 8-Pette™ and 12-Pette™ | |
Pipette |
8 | |
Non-sterile | |
20 to 200 μL |