Learn More
Corning™ Costar™ 8-Pette™ and 12-Pette™ Multichannel Pipetters
Adjustable volume pipetters can use any number of channels without affecting pipetting accuracy
£1209.14 - £1314.18
Specifications
Autoclavable Autoclavable
Product Line Costar™ 8-Pette™ and 12-Pette™
Sterility Non-sterile
Type Pipette
Volume (Metric) 20 to 200 μL
View More Specs
Products 2
Product Code Brand Number of Channels Price Quantity & Availability  
Product Code Brand Number of Channels Price Quantity & Availability  
10417611
View Documents
Corning™
4880
12 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10419521
View Documents
Corning™
4888
8 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
These20-200 μL multi-channel pipettors feature a unique, ergonomic trigger-style aspiration and dispense control mechanism designed to reduce thumb fatigue during repetitive pipetting.
• Volume adjusts with a vernier-scale spindle
• Pipettors are entirely autoclavable
Order Info
Compatible with all Fisherbrand™ universal-fit tips.
Specifications
Specifications
Autoclavable
Non-sterile
20 to 200 μL
Costar™ 8-Pette™ and 12-Pette™
Pipette