Learn More
Corning™ Thermowell™ 96-Well Polypropylene PCR Microplates
Constructed of medical grade polypropylene to accommodate wide temperature changes that might occur from storage to thermal cycling. Corning™ Thermowell™ 96-Well Polypropylene PCR Microplates are ideal for the polyerase chain reaction (PCR) method of DNA amplification.
Brand:  Corning™ 6551
Code : 66
Additional Details : Weight : 0.90718kg
 View more versions of this product
Product Code. 10727811
  • £172.37 / Pack of 25
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
Corning Thermowell GOLD 96-well PCR microplates are offered in four formats to ensure maximum flexibility and a perfect match for your applications. The original Thermowell 96-well PCR microplates are universal fit and can be cut into 3 x 8 well segments.
Specifications
Specifications
No Barcode
Standard
Can be cut into 3 x 8 well strips for lower throughput applications. Available PCR plate adapter for use with ABI PRISM™ 3700 DNA Analyzer
Natural
96
200 μL
PCR
25/Pk.
DNase and RNase-free
Without Lid
8 x 12
Polypropylene
Non-sterile
Conical Bottom (V-well)
25/Cs.
25/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products