Learn More
Corning™ 96-Well Clear PVC Assay Microplates
Manufactured from clear flexible Polyvinyl Chloride (PVC) for economy and visibility. Corning™ 96-Well Clear PVC Assay Microplates present an excellent solution for laboratory miniaturization. Due to their flexible nature, these microplates are not compatible with automation.
Brand: Corning™ 2797
Code : 66
Additional Details : Weight : 1.73250kg
Description
Specifications
Standard | |
8 x 12 | |
PVC | |
50 to 150 μL | |
Non-treated | |
Solution-based assays, serial dilutions and general storage applications | |
25/Pk. |
Without Lid | |
Clear | |
96 | |
Non-sterile | |
Round | |
100/Cs. | |
100/Cs. |