missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Clear Multiple Well Plates

Untreated surface is hydrophobic in nature and binds bio-olecules through passive interactions. Corning™ Costar™ Clear Multiple Well Plates provide an ideal solution for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.

Brand:  Corning™ 3738

Code : 66

Additional Details : Weight : 6.46650kg

 View more versions of this product

Product Code. 11309504

  • £266.84 / Pack of 100
Estimated Shipment: 26-09-2024
to see stock.
Add to basket

Description

Description

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
Specifications

Specifications

Standard
1.9 cm2
Medium Binding
Polystyrene
24
Sterile
Flat Bottom (F-well)
5/Pk.
With Lid
6 x 4
Clear
3.4 mL
0.38 to 0.57 mL
Non-treated
100/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers