Learn More
Corning™ Costar™ Netwell™ Carrier Kit, Non-Sterile
GREENER_CHOICE
Allows simultaneous processing
Brand:  Corning™ 3521
 View more versions of this product
Product Code. 10515531
  • £159.00 / Pack of 8
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • These specially designed Netwell carriers and handles allow simultaneous processing of up to 12 Netwell inserts per carrier
  • Each carrier kit contains eight carriers and eight handles
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Carrier Kit
    24 mm
    8/Cs.
    Allows simultaneous processing of up to 6 samples per carrier
    8/Pk.
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Product Information
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Corning™ Costar™ Netwell™ Carrier Kit, Non-Sterile > Carrier Kit, Diameter: 24mm