missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Transwell™-COL Collagen-Coated PTFE Membrane Insert
GREENER_CHOICE

Transparent, collagen-treated microporous membranes promote cell attachment and spreading

Brand:  Corning™ 3496

 View more versions of this product

Product Code. 10377791

This item is not returnable. View return policy

Description

Description

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

You can use cell culture inserts to:

  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings Transparent, Collagen-coated PTFE membrane
  • Promotes cell attachment and spreading
  • Equimolar mixture of Collagen type I and III
  • Individually packaged
  • Multiple well plates included in each case
  • Sterile

Order Info

Individually packed and supplied in package with tissue culture plates with lids; sterile.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.33 cm2
0.6 mL
Membrane Insert
6.5 mm
24/Cs.
Sterile
PTFE
3 μm
24 Well Plate
1/Pk.
Collagen
24
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers