missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Transwell™ Inserts and Dishes

Applications include co-culture, drug transport, in vitro fertilization, and microbial pathogenesis studies.

Brand:  Corning™ 3420

Code : 67

Additional Details : Weight : 0.58967kg

 View more versions of this product

Product Code. 10164381

  • £612.80 / Pack of 12
Estimated Shipment: 11-12-2024
to see stock.
Add to basket

Description

Description

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

You can use cell culture inserts to:

  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 μm thick translucent membrane
  • Pore sizes ranging from 0.4 μm to 8 μm diameters
  • Treated for optimal cell attachment
  • Supplied in multiple well plates
  • Membrane must be stained for cell visibility
  • Sterile
Specifications

Specifications

Round
Clear
Polycarbonate
Cell Culture
9 mL
1/Pk.
3.0 μm
13 mL (Well)
44 cm2
Sterile
Cell Culture Insert
12/Cs.
3 μm
75 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers