missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Costar™ Transwell™ Inserts and Dishes
Applications include co-culture, drug transport, in vitro fertilization, and microbial pathogenesis studies.
£526.46 - £612.80
Specifications
Bottom Shape | Round |
---|---|
Culture Area | 44 cm2 |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Type | Cell Culture Insert |
Product Code | Brand | Supportive Membrane | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Supportive Membrane | Price | Quantity & Availability | |||||
10164381
|
Corning™
3420 |
3.0 μm |
£612.80
Pack of 12 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
10630312
|
Corning™
3419 |
0.4 μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
You can use cell culture inserts to:
- Produce a cell culture environment that closely resembles an in vivo state
- Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
- Co-culture cells with or without cell-to-cell contact
- Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 μm thick translucent membrane
- Pore sizes ranging from 0.4 μm to 8 μm diameters
- Treated for optimal cell attachment
- Supplied in multiple well plates
- Membrane must be stained for cell visibility
- Sterile
Order Info
Individually wrapped. Twelve inserts (with dishes) per case.
Specifications
Round | |
Sterile | |
Cell Culture Insert | |
75 mm |
44 cm2 | |
Cell Culture | |
9 mL |