missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Biohazard Autoclave Gloves
Shop All Cole-Parmer Products
Click to view available options
Color:
Orange
Description
- Heat resistant to 450°F (232°C)
- Excellent for use in any environment where heat protection is required
- Bright orange for increased visibility
- Machine wash, tumble dry
Specifications
Specifications
Cuff Material | Cotton |
Color | Orange |
Product Type | Gloves |
Size | Universal |
Quantity | 1 Pack |
Resistant To | Heat |
Length (Metric) | 330 mm |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Biohazard Autoclave Gloves >
Spot an opportunity for improvement?Share a Content Correction