missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ All Cotton\Terry Autoclave gloves
Shop All SP Scienceware Products
Click to view available options
Length (Metric):
330 mm
Color:
Orange
Glove Cuff Style:
Gauntlet
Temperature (Metric) Max:
232°C
Description
For handling hot objects out of the autoclave or oven, or for other heat protection. Bel Art™ All Cotton\Terry Autoclave gloves are soft and pliant yet durable.
- Heat-resistant up to 232°C; replace hazardous asbestos gloves
- Gauntlets in two lengths provide wrist and forearm protection
- Bright orange color offers high visibility
- Machine wash, air dry, or use the Lab-Aire™ II Glove Holder
- CE Marked
- NOT waterproof
- Do NOT use with dry ice
Specifications
Specifications
Color | Orange |
Laboratory/Cleanroom Use | Yes |
Latex Free | Yes |
Material | Cotton, Terrycloth |
Powdered | Powder Free |
Product Type | Gloves |
Temperature (Metric) Max | 232°C |
Glove Cuff Style | Gauntlet |
Quantity | 1 Ea. |
Resistant To | Heat |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel-Art™ All Cotton\Terry Autoclave gloves >
Spot an opportunity for improvement?Share a Content Correction