missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Density | 1.070g/cm3 |
---|---|
Flash Point | 11°C (51.8°F) |
Chemical Name or Material | HYDRANAL™-Coulomat A |
Boiling Point | 59°C |
Packaging | Polycoated bottle |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15684450
|
Honeywell Fluka
34807-500ML |
500 mL | Polycoated bottle |
£96.56
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15694450
|
Honeywell Fluka
34807-6X500ML |
6 x 500 mL | Polycoated bottle |
£504.31
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Specifications
1.070g/cm3 | |
HYDRANAL™-Coulomat A | |
Polycoated bottle |
11°C (51.8°F) | |
59°C | |
UN3286 |
Safety and Handling
P201-P210-P280-P308 + P311-P370 + P378-P403 + P235
missing translation for 'dotInformation' : 8(6.1)(3)/PGII
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL-Coulomat A
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.,Suspected of causing cancer.,Suspected of causing cancer.
- H331-Toxic if inhaled.
- H360D-May damage the unborn child.,May damage the unborn child.,May damage the unborn child.
- H370-Causes damage to organs.,Causes damage to organs.,Causes damage to organs.
- H336-May cause drowsiness or dizziness.
- H372-Causes damage to organs.,Causes damage to organs.,Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : trichloromethane; chloroform, methanol,imidazole, sulphur dioxide, 1H-Imidazolemonohydriodide