Learn More
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals
£35.88 - £443.59
Chemical Identifiers
CAS | 531-81-7 |
---|---|
Molecular Formula | C10H6O4 |
Molecular Weight (g/mol) | 190.15 |
MDL Number | MFCD00006852 |
InChI Key | ACMLKANOGIVEPB-UHFFFAOYSA-N |
Synonym | coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 van |
PubChem CID | 10752 |
IUPAC Name | 2-oxochromene-3-carboxylic acid |
SMILES | C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10525061
|
Thermo Scientific Acros
346760250 |
25 g | Glass bottle |
£35.88
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10295671
|
Thermo Scientific Acros
346761000 |
100 g | Glass bottle |
£110.55
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10239593
|
Thermo Scientific Acros
346765000 |
500 g | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
531-81-7 | |
190.15 | |
ACMLKANOGIVEPB-UHFFFAOYSA-N | |
10752 | |
C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O |
C10H6O4 | |
MFCD00006852 | |
coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 van | |
2-oxochromene-3-carboxylic acid |
Specifications
531-81-7 | |
Brown | |
98% | |
MFCD00006852 | |
15, 2551 | |
Solubility in water: 13g/L (37°C). Other solubilities: soluble in alcohol and alkalies,insoluble in ether,benzene,petroleum ether | |
C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O | |
190.15 | |
190.15 | |
Crystalline Powder |
190°C to 193°C | |
Authentic | |
C10H6O4 | |
18, 429 | |
coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 van | |
ACMLKANOGIVEPB-UHFFFAOYSA-N | |
2-oxochromene-3-carboxylic acid | |
10752 | |
98% | |
Coumarin-3-carboxylic acid |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-518-
RUO – Research Use Only