missing translation for 'onlineSavingsMsg'
Learn More
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 11337156

Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11337156
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11337156

Brand: Thermo Scientific Alfa Aesar L07133.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Coumarin-3-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coumarin-3-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Soluble in water (13 g/L at 37°C) and alcohol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-81-7
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00006852
InChI Key ACMLKANOGIVEPB-UHFFFAOYSA-N
Synonym coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 vanShow MoreShow Less
PubChem CID 10752
IUPAC Name 2-oxochromene-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O

Specifications

Melting Point 190°C to 193°C
Quantity 100 g
UN Number UN2811
Beilstein 154276
Merck Index 14,2563
Solubility Information Soluble in water (13g/L at 37°C) and alcohol.
Formula Weight 190.15
Percent Purity 98%
Chemical Name or Material Coumarin-3-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453229 Certificate TypeCertificate of Analysis Date18/10/2023 Catalog NumberL07133.22,L07133.14
Lot NumberA0449131 Certificate TypeCertificate of Analysis Date18/10/2023 Catalog NumberL07133.22,L07133.14
Lot Number10210562 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL07133.22,L07133.14
Lot Number10236720 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL07133.22,L07133.14
Lot Number10236860 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL07133.22,L07133.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.