Learn More
Cole-Parmer™ CPC™ Panel Mount Gasket Couplers
Virtually eliminate any loss of product with this leak-free design
£63.55 - £64.15
Specifications
Type Panel Mount Gasket
Packaging Quantity 10 Gaskets
Material EPDM
View More Specs
Products 2
Product Code Brand For Use With Price Quantity & Availability  
Product Code Brand For Use With Price Quantity & Availability  
15427333
View Documents Promotion Details
Cole-Parmer™
31603-18
NS4 Series Couplers
£64.15
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
15437333
View Documents Promotion Details
Cole-Parmer™
31603-19
NS6 Series Couplers This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance.
  • For a complete fitting, order one body and one insert.
  • All bodies will mate with inserts to make a fitting that will work for multiple applications.
Specifications
Specifications
Panel Mount Gasket
10 Gaskets
EPDM