Learn More
Cole-Parmer™ CPC™ Quick Disconnect Polypropylene Pipe Adapter Bodies
Quick Disconnect couplings feature a thumb latch for easy assembly
Brand:  Cole-Parmer™ 31303-07
 View more versions of this product
Product Code. 15447053
  • £20.70 / Each
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High-flow quick-disconnect couplings consists of a body and an insert (order fitting halves separately)
  • Plug insert into coupling body to connect
  • Fitting halves lock into place immediately
  • To disconnect fitting halves, simply press the button on the coupling body
  • Valved models have a spring-operated 316 SS valve that seals the flow path when fitting halves are disengaged
  • All fitting bodies have 316 SS latch springs and EPDM seals
Specifications
Specifications
Adapter
71°C
3/4 in. Quick-Disconnect
Ethylene Oxide (ETO) or Gamma
60 psi
160°F
3/4 in.
Quick-Disconnect x NPT(M) Connection
Polypropylene