Learn More
Cole-Parmer™ CPC™ Quick-Disconnect Valved Coupling Insert
Quick-disconnect fitting with simple push-to-connect interface for your rigid tubing
Brand:  Cole-Parmer™ 31304-32
 View more versions of this product
Product Code. 11745358
  • £25.80 / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Plug insert into body to connect fitting.
  • Fitting halves lock in place immediately.
  • To disconnect fitting halves, simply press the thumb latch on the coupling body.
  • spring-operated valve in each fitting half seals the flow path.
  • Straight-through fittings without shutoff valves are also available.
  • All fitting bodies have 316 stainless steel springs and latches.
Specifications
Specifications
Quick-Disconnect Valved Coupling Insert
180°F
Quick-Disconnect x Push-to-Connect
Autoclave or Ethylene Oxide (ETO)
Acetal
White
82°C
1/4 x 3/8 in.
4.98 cm
120 psi