missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CpG oligodeoxynucleotides (2216), Novus Biologicals™
Useful positive control model system for studying hTLR9 activation
£131.75 - £191.45
Specifications
Concentration | 1.0 mg/mL |
---|---|
Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Type | CpG Oligodeoxynucleotides |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18015644
|
Bio-Techne
NBP2-31136 |
0.1 mg |
£191.45
0.10mg |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18154912
|
Bio-Techne
NBP2-31136SS |
0.025 mg |
£131.75
25µg |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Immunogen: G*GGGGACGATCGTCG*G*G*G*G*GBuffer: Sterile water
Clone: 2216
Dilution: Ligand Activation, 5 to 20μg/mL
CpG ODN Type A (2216)
Specifications
1.0 mg/mL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
CpG Oligodeoxynucleotides | |
Human |