missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CpG oligodeoxynucleotides Mouse

Applications: Ligand Activation

Brand:  Novus Biologicals™ NBP2-31132SS

Product Code. 18178333

  • £135.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
0.025 mg
Activation of mouse TLR9
CpG Oligodeoxynucleotides
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers