Learn More
Novus Biologicals™ CpG oligodeoxynucleotides Mouse
Brand:  Novus Biologicals™ NBP2-31142
Product Code. 18046855
  • £193.00 / 0.10mg
Explore more special offers
Specifications
1.0 mg/mL
0.1 mg
CpG Oligodeoxynucleotides Mouse
Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Mouse CpG ODNs Are Used for Activation of Mouse TLR9 (at a Range of 5-20 μg/mL The TLR9/NF-kB/SEAPorter HEK 293 Cell Line is a Useful Positive Control Model System for Studying hTLR9 Activation. We Typically Use a 96-Well Plate Format Assay Using This Reporter Cell Line.)
CpG Oligodeoxynucleotides
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Novus Biologicals™ CpG oligodeoxynucleotides Mouse >