Learn More
Novus Biologicals™ CpG oligodeoxynucleotides Mouse
Brand: Novus Biologicals™ NBP2-31140SS
Additional Details : Weight : 0.00970kg
Specifications
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
0.025 mg | |
Activation of mouse TLR9 | |
CpG Oligodeoxynucleotides |