Learn More
WTW™ Crack Set
Brand: WTW™ 252033
145.90 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description

Specifications
Crack Set | |
10 L |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.