Learn More
Abeba™ Crawler ALU 34581 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Brand:  Abeba™ 34581/42
Additional Details : Weight : 1.21500kg
 View more versions of this product
Product Code. 15650028
  • £125.43 / 1 pair
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • CE EN ISO 20345:2011 S1 SRC
  • Velours
  • Black/Blue
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRC
Velour
Slip-resistant
34581
Black, Blue
42
Polyurethane
Aluminum