Learn More
CellSensor™ CRE HEK 293T DA Assay Kit
CellSensor™ CRE HEK 293T DA Assay Kit contains a beta-lactamase (bla) reporter gene under control of a cAMP response element (CRE).
Brand:  CellSensor™ K1457
Code : NEW
Additional Details : Weight : 0.05000kg
Product Code. 10206844
  • £644.00 / Each
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
Description
Description
  • Fluorescence activated cell sorting (FACS) was used to isolate clones responsive to stimulation of the CRE pathway by forskolin. The cell line was validated for DMSO tolerance, cell number, stimulation time, substrate loading conditions, and EC50 concentration of forskolin. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
  • This product can serve as a negative control in screening assays performed with specific G-protein coupled receptor (GPCR) DA cells that were made from this same background. These cells can be used in other assays where a sensitive readout to intracellular changes to cAMP is needed.
Specifications
Specifications
Should be stored in liquid nitrogen immediately upon receipt
CellSensor™ CRE HEK293T DA cells, LiveBLAzer™-FRET B/G Loading Kit 70 μg, Solution D, CellSensor™ CRE HEK293T CHO-K1 Assay Protocol, LiveBLAzer™-FRET B/G Loading Protocol, Certificate of Analysis
Division-Arrested Cells Assay Kit
GPCRs, Signaling Pathway
1 Plate