missing translation for 'onlineSavingsMsg'
Learn More

Creatine phosphate disodium salt tetrahydrate, 99%, Thermo Scientific Chemicals

£24.32 - £131.66

Chemical Identifiers

CAS 71519-72-7
Molecular Formula C4H8N3Na2O5P
Molecular Weight (g/mol) 255.08
MDL Number MFCD00150192
InChI Key RNTXMYSPASRLFT-UHFFFAOYSA-L
Synonym creatine phosphate sodium salt
PubChem CID 91884830
SMILES [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10579263
View Documents
Thermo Scientific Acros
337340010
1 g Glass bottle
£24.32
1g
Please to purchase this item. Need a web account? Register with us today!
10616794
View Documents
Thermo Scientific Acros
337340020
2 g Glass bottle
£39.40
2g
Please to purchase this item. Need a web account? Register with us today!
10216011
View Documents Promotion Details
Thermo Scientific Acros
337340100
10 g Glass bottle
£131.66
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

71519-72-7
255.08
RNTXMYSPASRLFT-UHFFFAOYSA-L
91884830
C4H8N3Na2O5P
MFCD00150192
creatine phosphate sodium salt
[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O

Specifications

71519-72-7
Authentic
C4H8N3Na2O5P
MFCD00150192
(10% in water) Clear colorless solution
[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
255.08
327.14
Powder
White
99%
Na2O3PNHC(NH)N(CH3)CH2COOH·4H2O
creatine phosphate sodium salt
RNTXMYSPASRLFT-UHFFFAOYSA-L
disodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
91884830
99%
Creatine phosphate disodium salt tetrahydrate
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Creatine phosphate disodium salt tetrahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only