missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CREB1 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FS0059.200uL
Additional Details : Weight : 0.02000kg
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
CREB,MGC9284 | |
1385 | |
200 μL |
Tube | |
cAMP responsive element binding protein 1 | |
CREB1 | |
DNA/RNA | |
Liquid |