missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CREB1 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
£1235.00 - £2030.00
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | cAMP responsive element binding protein 1 |
Gene Alias | CREB,MGC9284 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043505
|
Abnova™
FS0059.100UL |
100 μL |
£1235.00
100µL |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053505
|
Abnova™
FS0059.200UL |
200 μL |
£2030.00
200µL |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
CREB,MGC9284 | |
1385 | |
Liquid |
Tube | |
cAMP responsive element binding protein 1 | |
CREB1 | |
DNA/RNA |