Learn More
Macherey-Nagel™ Crimping Head
Brand:  Macherey-Nagel™ 735711
 View more versions of this product
Product Code. 12888595
  • £383.00 / Each
Estimated Shipment: 29-10-2024
to see stock.
Explore more special offers
Add to basket
Specifications
Crimping Head
1 Each
For Electronic High Power Crimping Tool 735700 and 11 mm Crimp Caps