missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Crimping Head
£383.00
Código de producto | Marca | For Use With (Equipment) | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | For Use With (Equipment) | Precio | Cantidad y disponibilidad | |||||
12888595
|
Macherey-Nagel™
735711 |
For Electronic High Power Crimping Tool 735700 and 11 mm Crimp Caps |
£383.00
Each |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
12898595
|
Macherey-Nagel™
735720 |
For Electronic High Power Crimping Tool 735700 and 20 mm Crimp Caps |
£383.00
Each |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
Especificaciones
Crimping Head | |
1 Each |