missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody (6G12) - C-terminus - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-52398SS
Additional Details : Weight : 0.00970kg
Description
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP).Specifications
CRISPR-Cas9 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:1000, Simple Western 10-20 ug/ml, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) | |
Cas9, CRISPR, CRISPR-associated protein 9 nuclease, SpyCas9 | |
Recombinant C-terminal fragment of S.pyogenes CRISPR/Cas9. [UniProt# Q99ZW2] | |
0.025 mL | |
Gene Editing, Gene Knockouts, Gene Therapy | |
Cas9 from Streptococcus pyogene. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay | |
6G12 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Bacteria | |
Purified |