missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody (6G12) - C-terminus - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£183.58 - £443.77
Specifications
Antigen | CRISPR-Cas9 |
---|---|
Clone | 6G12 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:1000, Simple Western 10-20 ug/ml, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18232560
|
Bio-Techne
NBP2-52398SS |
0.025 mL |
£183.58
25µL |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18095476
|
Bio-Techne
NBP2-52398 |
0.1 mL |
£443.77
0.10mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP).Specifications
CRISPR-Cas9 | |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, ChIP Assay | |
Unconjugated | |
Mouse | |
Gene Editing, Gene Knockouts, Gene Therapy | |
PBS with 0.02% Sodium Azide | |
Recombinant C-terminal fragment of S.pyogenes CRISPR/Cas9. [UniProt# Q99ZW2] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
6G12 | |
Western Blot 1:1000, Simple Western 10-20 ug/ml, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) | |
Monoclonal | |
Purified | |
RUO | |
Bacteria | |
Cas9, CRISPR, CRISPR-associated protein 9 nuclease, SpyCas9 | |
IgG1 κ | |
Protein G purified | |
Cas9 from Streptococcus pyogene. |