Learn More
Cromolyn sodium salt, 98%, Thermo Scientific Chemicals
Mast cell stabilizer that prevents release of histamine from mast cells
£42.51 - £412.05
Chemical Identifiers
CAS | 15826-37-6 |
---|---|
Molecular Formula | C23H14Na2O11 |
Molecular Weight (g/mol) | 512.33 |
MDL Number | MFCD00057744 |
InChI Key | VLARUOGDXDTHEH-UHFFFAOYSA-L |
Synonym | cromolyn sodium, sodium cromoglycate, disodium cromoglycate, sodium cromoglicate, sodium cromolyn, gastrocrom, nasalcrom, cromoglycate disodium, natrium cromoglicat, lomudal |
PubChem CID | 27503 |
ChEBI | CHEBI:128458 |
SMILES | [Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15436739
|
Thermo Scientific Alfa Aesar
J65122.03 |
1 g |
£42.51
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15446739
|
Thermo Scientific Alfa Aesar
J65122.06 |
5 g |
£159.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15456739
|
Thermo Scientific Alfa Aesar
J65122.14 |
25 g |
£412.05
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cromolyn sodium salt is used as a mast cell stabilizer that prevents release of histamine and other pro-inflammatory mediators from mast cells. Cromolyn sodium salt is used as a chemopreventive and an anti-inflammatory agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCromolyn sodium salt is used as a mast cell stabilizer that prevents release of histamine and other pro-inflammatory mediators from mast cells. Cromolyn sodium salt is used as a chemopreventive and an anti-inflammatory agent.
Solubility
Soluble in water to 100mg/ml
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
15826-37-6 | |
512.33 | |
VLARUOGDXDTHEH-UHFFFAOYSA-L | |
27503 | |
[Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O |
C23H14Na2O11 | |
MFCD00057744 | |
cromolyn sodium, sodium cromoglycate, disodium cromoglycate, sodium cromoglicate, sodium cromolyn, gastrocrom, nasalcrom, cromoglycate disodium, natrium cromoglicat, lomudal | |
CHEBI:128458 |
Specifications
15826-37-6 | |
C23H14Na2O11 | |
14,2590 | |
Soluble in water to 100mg/ml | |
[Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O | |
512.33 | |
CHEBI:128458 | |
98% | |
Cromolyn sodium salt |
White to Yellow | |
MFCD00057744 | |
cromolyn sodium, sodium cromoglycate, disodium cromoglycate, sodium cromoglicate, sodium cromolyn, gastrocrom, nasalcrom, cromoglycate disodium, natrium cromoglicat, lomudal | |
VLARUOGDXDTHEH-UHFFFAOYSA-L | |
disodium;5-[3-(2-carboxylato-4-oxochromen-5-yl)oxy-2-hydroxypropoxy]-4-oxochromene-2-carboxylate | |
27503 | |
512.33 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 239-926-7
missing translation for 'rtecsNumber' : DJ2380000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.