Learn More
Welch™ CRVpro™ Starter Package
The robust vacuum pump CRVpro belongs to the new high-performance two-stage rotary vane pumps generation of Welch.
£3360.20 - £3380.00
Specifications
Includes CRVpro with Oil Mist Separator AKD 16, 2-Way Ball Valve, Hose Connector, Vacuum Hose (1.5 m), Oil Change Set
Pump Type Rotary Vane Vacuum Pump
View More Specs
Products 2
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
15721918
View Documents
Welch™
330071
CRVpro 4
£3360.20
Each
Estimated Shipment: 30-07-2025
to see stock.
Add to basket
15716809
View Documents Promotion Details
Welch™
330073
CRVpro 8
£3380.00
Each
Estimated Shipment: 30-07-2025
to see stock.
Add to basket
Description
Description
  • CRVpro Starter Package Includes:
    • Oil mist separator AKD 16
    • 2-way ball valve
    • Hose connector DN 16KF-10/8 mm
    • Vacuum hose (1.5 m)
    • Oil change set
Specifications
Specifications
CRVpro with Oil Mist Separator AKD 16, 2-Way Ball Valve, Hose Connector, Vacuum Hose (1.5 m), Oil Change Set
Rotary Vane Vacuum Pump
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Welch™ CRVpro™ Starter Package