Learn More
Tempshield™ Cryo-Gloves™ Blue Waterproof Wrist-Length Cryogenic Gloves
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
Brand: Tempshield™ WRMWP
277.69 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Description
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
- Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
- Thin, seamless 100% waterproof liner provides protection from spills and splashes
- State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
- High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
- Ideal for applications where function is important and safety is critical
- Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
EN 420, EN 511 | |
Blue | |
Waterproof Cryogenic Gloves | |
Yes | |
Cryo-Gloves™ | |
-196°C | |
28 to 33 cm |
Wrist Length | |
Specific | |
Medium | |
Dispensing/Transferring Cryogenic Liquids, Removing Samples from Cryogenic Liquids, and Open Cryogenic Containers with the Chance of Exposure to Liquids/Splashes | |
-320°F | |
11 to 13 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.