missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tempshield™ Cryo-Gloves™ Blue Waterproof Wrist-Length Cryogenic Gloves
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
£323.00 - £333.34
Specifications
Certifications/Compliance (Europe) | EN 420, EN 511 |
---|---|
Description | Wrist Length |
Color | Blue |
Hand | Specific |
Product Type | Waterproof Cryogenic Gloves |
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
13453090
|
Tempshield™
WRSWP |
Small |
£323.00
1 pair |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||||
11863831
|
Tempshield™
WRMWP |
Medium |
£323.00
1 pair |
In Stock
Log in to see stock. |
|||||
11873831
|
Tempshield™
WRLWP |
Large |
£333.34
1 pair |
In Stock
Log in to see stock. |
|||||
Description
A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
- Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
- Thin, seamless 100% waterproof liner provides protection from spills and splashes
- State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
- High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
- Ideal for applications where function is important and safety is critical
- Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
EN 420, EN 511 | |
Blue | |
Waterproof Cryogenic Gloves | |
Dispensing/Transferring Cryogenic Liquids, Removing Samples from Cryogenic Liquids, and Open Cryogenic Containers with the Chance of Exposure to Liquids/Splashes | |
-320°F | |
11 to 13 in. |
Wrist Length | |
Specific | |
Yes | |
Cryo-Gloves™ | |
-196°C | |
28 to 33 cm |