Learn More
Thermo Scientific™ CryoBox™ Freezer Racks
GREENER_CHOICE
Stack and organize standard cryogenic vial storage boxes with these freezer racks.
Brand:  Thermo Scientific™ DS5037-0007
304.41 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 10718861
  • £422.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Stack and organize standard cryogenic vial storage boxes with these freezer racks. Thermo Scientific™ CryoBox™ Freezer Racks include durable Thermo Scientific Nalgene™ Stainless Steel CryoBox Racks and Thermo Scientific Nunc™ aluminum racks that fit most common upright and chest freezers.
  • Conveniently store standard 5-1/4 x 5-1/4 cryogenic vial storage boxes with these freezer racks
  • Fits Thermo Scientific Nalgene™ and Nunc™ CryoBoxes™
  • Options include durable stainless-steel and aluminum racks that fit most common upright and chest freezers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Silver
7 CryoBoxes/Rack
Stainless Steel
5.625 x 5.5 x 27.75 in. (14.3 x 14 x 70.5 cm)
Vertical
Freezer Rack