Learn More
Thermo Scientific™ CryoPlus™ Series Racks and Holders
Increase the usability of Thermo Scientific™ CryoPlus™ LN2 storage systems with CryoPlus Series Racks and Holders.
£218.68 - £1101.87
Product Code | Brand | Type | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | Price | Quantity & Availability | |||||
10294604
|
Thermo Scientific™
4000001 |
Square Rack, Vapor Phase | 11 Cardboard Boxes (100 Cell Divider) |
£391.59
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15186265
|
Thermo Scientific™
4000002 |
Square Rack, Vapor Phase | 11 Stainless Steel Boxes (100 Cell Divider) |
£762.84
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
10355914
|
Thermo Scientific™
4000003 |
Square Rack, Vapor Phase | 7 Cardboard Boxes (100 Cell Divider) |
£317.00
Each |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||||
10011684
|
Thermo Scientific™
4000005 |
Square Rack, Vapor Phase | 6 Cardboard Boxes (100 Cell Divider) |
£244.10
Each |
Estimated Shipment: 15-08-2025
Log in to see stock. |
|||||
10315964
|
Thermo Scientific™
4000006 |
Square Rack, Liquid Phase | 13 Cardboard Boxes (100 Cell Divider) |
£465.33
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
11894540
|
Thermo Scientific™
4000007 |
Square Rack, Liquid Phase | 13 Stainless Steel Boxes (100 Cell Divider) |
£713.67
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||
10254564
|
Thermo Scientific™
4000008 |
Square Rack, Liquid Phase | 8 Cardboard Boxes (100 Cell Divider) |
£361.92
Each |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||||
11786509
|
Thermo Scientific™
4000010 |
Square Rack, Liquid Phase | 7 Cardboard Boxes (100 Cell Divider) |
£304.28
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||||
10795715
|
Thermo Scientific™
4000042 |
Square Rack, Liquid Phase | 12 Cardboard Boxes (100 Cell Divider) |
£542.46
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
10376174
|
Thermo Scientific™
4000043 |
Arrowhead Rack, Liquid Phase | 7 Cardboard Boxes (81 Cell Divider) |
£540.76
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
10386174
|
Thermo Scientific™
4000044 |
Arrowhead Rack, Liquid Phase | 13 Cardboard Boxes (81 Cell Divider) |
£795.05
Each |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||||
10375764
|
Thermo Scientific™
4000108 |
Arrowhead Rack, Liquid Phase | 10 Cardboard Boxes (81 Cell Divider) |
£590.77
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||||
15531724
|
Thermo Scientific™
4000111 |
Arrowhead Rack, Vapor Phase | 11 Stainless Steel Boxes (81 Cell Divider) |
£1101.87
Each |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||||
11834770
|
Thermo Scientific™
4000112 |
Arrowhead Rack, Liquid Phase | 12 Cardboard Boxes (81 Cell Divider) |
£756.90
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||||
15742522
|
Thermo Scientific™
4000113 |
Arrowhead Rack, Liquid Phase | 12 Stainless Steel Boxes (81 Cell Divider) |
£590.77
Each |
Estimated Shipment: 12-08-2025
Log in to see stock. |
|||||
10264564
|
Thermo Scientific™
4000011 |
Vertical Rack, Vapor Phase | 4 Boxes |
£218.68
Each |
Estimated Shipment: 09-10-2025
Log in to see stock. |
|||||
10506474
|
Thermo Scientific™
4000012/99283 |
Vertical Rack, Liquid Phase | 5 Cardboard Boxes (100 Cell Divider) |
£277.16
Each |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||||
Description
Increase the usability of Thermo Scientific™ CryoPlus™ LN2 storage systems with CryoPlus Series Racks and Holders.
Vertical Racks, Liquid Phase
- 2mL, 5 cardboard boxes (100 cell divider)
- 2mL, 12 carboard boxes (25 cell divider)
Vertical Racks, Vapor Phase
- 2mL, 4 cardboard boxes (100 cell divider)
Square Racks, Liquid Phase
- 2mL, 12 cardboard boxes (100 cell divider)
- 2mL, 13 cardboard boxes (100 cell divider)
- 2mL, 13 stainless steel boxes (100 cell divider)
- 4mL, 8 cardboard boxes (100 cell divider)
- 4mL, 8 stainless steel boxes (100 cell divider)
- 5mL, 7 cardboard boxes (100 cell divider)
Square Racks, Vapor Phase
- 2mL, 11 cardboard boxes (100 cell divider)
- 2mL, 11 stainless steel boxes (100 cell divider)
- 4mL, 7 cardboard boxes (100 cell divider)
- 4mL, 7 stainless steel boxes (100 cell divider)
- 5mL, 6 cardboard boxes (100 cell divider)
Canister Cane Storage, Vapor Phase
- 2mL, 4mL, 5mL vial and straw canes, includes 8 canisters
Arrowhead Racks, Liquid Phase
- 2mL, 12 cardboard boxes (81 cell divider)
- 2mL, 13 cardboard boxes (81 cell divider)
- 2mL, 12 stainless steel boxes (81 cell divider)
- 4mL, 8 cardboard boxes (81 cell divider)
- 4mL, 8 stainless steel boxes (81 cell divider)
- 5mL, 7 cardboard boxes (81 cell divider)
Arrowhead Racks, Vapor Phase
- 2mL, 11 cardboard boxes (81 cell divider)
- 2mL, 11 stainless steel boxes (81 cell divider)
- 2mL, 10 cardboard boxes (81 cell divider)
- Warranty varies by country, please contact us for details