Learn More
STEMCELL Technologies CryoStor™ CS10
Animal-component-free, defined cryopreservation medium with 10% DMSO
Brand:  STEMCELL Technologies 07955
Encompass_Preferred
 View more versions of this product
Product Code. 17159931
  • £643.95 / 100mL
Explore more special offers
Description
Description
  • CryoStor™ CS10 is a uniquely formulated serum-free, animal component-free, and defined cryopreservation medium containing 10% dimethyl sulfoxide (DMSO)
  • Designed to preserve cells in low temperature environments (-80°C to -196°C), CryoStor™ CS10 provides a safe, protective environment for cells and tissues during the freezing and thawing processes and during storage
  • CryoStor™ CS10 is recommended for the cryopreservation of hepatocytes, tissue samples, human peripheral blood, CHO cells, myeloma cell lines, hybridomas, human mesenchymal stem cells, human embryonic and human induced pluripotent stem cells (ES cells and iPS cells) and other extremely sensitive cell types
  • CryoStor™ CS10 is cGMP-manufactured with USP grade components
Specifications
Specifications
B Cells, CHO Cells, Hematopoietic Stem and Progenitor Cells, Hybridomas, Intestinal Cells, Macrophages, Mesenchymal Stem and Progenitor Cells, Monocytes, Myeloma, NK Cells, Other, Pluripotent Stem Cells, T Cells
Cryopreservation
100 mL Bag