missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies CryoStor™ CS10
Animal-component-free, defined cryopreservation medium with 10% DMSO
£394.20 - £6562.35
Specifications
Cell Type | B Cells, CHO Cells, Hematopoietic Stem and Progenitor Cells, Hybridomas, Intestinal Cells, Macrophages, Mesenchymal Stem and Progenitor Cells, Monocytes, Myeloma, NK Cells, Other, Pluripotent Stem Cells, T Cells |
---|---|
For Use With (Application) | Cryopreservation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15642424
|
STEMCELL Technologies
07931 |
5 x 16 mL |
£511.65
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17159931
|
STEMCELL Technologies
07955 |
100 mL Bag |
£643.95
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17169931
|
STEMCELL Technologies
07959 |
5 x 10 mL |
£394.20
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17199921
|
STEMCELL Technologies
07940 |
1000 mL Bag |
£6562.35
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- CryoStor™ CS10 is a uniquely formulated serum-free, animal component-free, and defined cryopreservation medium containing 10% dimethyl sulfoxide (DMSO)
- Designed to preserve cells in low temperature environments (-80°C to -196°C), CryoStor™ CS10 provides a safe, protective environment for cells and tissues during the freezing and thawing processes and during storage
- CryoStor™ CS10 is recommended for the cryopreservation of hepatocytes, tissue samples, human peripheral blood, CHO cells, myeloma cell lines, hybridomas, human mesenchymal stem cells, human embryonic and human induced pluripotent stem cells (ES cells and iPS cells) and other extremely sensitive cell types
- CryoStor™ CS10 is cGMP-manufactured with USP grade components
Specifications
B Cells, CHO Cells, Hematopoietic Stem and Progenitor Cells, Hybridomas, Intestinal Cells, Macrophages, Mesenchymal Stem and Progenitor Cells, Monocytes, Myeloma, NK Cells, Other, Pluripotent Stem Cells, T Cells | |
Cryopreservation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title