Learn More
STEMCELL Technologies CryoStor™ CSB
Animal component-free and protein-free cryopreservation formulation
Brand:  STEMCELL Technologies 100-0238
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 17147501
  • £3692.25 / 500mL
Estimated Shipment: 23-07-2024
to see stock.
Description
Description
  • CryoStor™ CSB is an intracellular-like base formulation from the CryoStor™ preservation media series
  • It is serum-free, animal component-free, and does not contain dimethyl sulfoxide (DMSO)
  • It is designed to improve post-thaw cell viability and function, and to reduce the level of cryopreservation-induced delayed-onset cell death
Specifications
Specifications
B Cells, CHO Cells, Hybridomas, Intestinal Cells, Macrophages, Mesenchymal Stem and Progenitor Cells, Monocytes, Myeloma, NK Cells, Other, Pluripotent Stem Cells, T Cells
Cryopreservation
500 mL